accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-32-2

IUPAC Name: N-benzylaniline
Synonyms: N-benzylaniline
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: GTWJETSWSUWSEJ-UHFFFAOYSA-N
SMILES: C(NC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)