Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Benzylaniline

Catalog No. 185493100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-549-3100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-549-3100G Supplier Sigma Aldrich Supplier No. 185493100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Benzylaniline

Specifications

CAS 103-32-2
Melting Point 35°C to 38°C (lit.)
Density 1.061 g/mL (at 25°C (literature))
Boiling Point 306°C to 307°C (lit.)
Molecular Formula C13H13N
Linear Formula C6H5CH2NHC6H5
MDL Number MFCD00003018
Quantity 250 mg
Synonym N-Phenylbenzylamine
Molecular Weight (g/mol) 183.25
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.