accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 103-45-7

IUPAC Name: 2-phenylethyl acetate
Synonyms: phenethyl acetate β-phenethyl acetate phenethyl acetate (natural) 2-phenethyl acetate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: MDHYEMXUFSJLGV-UHFFFAOYSA-N
SMILES: CC(=O)OCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)