Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Phenethyl acetate

Catalog No. 2905805ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-058-05ML 5 mL
29-058-0250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-058-05ML Supplier Sigma Aldrich Supplier No. 2905805ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Phenethyl acetate is a highly valued natural volatile ester with a rose-like odour and is widely used to add scent or flavour to cosmetics, soaps, foods and drinks.

Specifications

CAS 103-45-7
Density 1.032 g/mL (at 25°C (literature))
Boiling Point 238°C to 239°C (lit.)
Molecular Formula C10H12O2
Refractive Index n20/D 1.498 (literature)
Linear Formula CH3COOCH2CH2C6H5
MDL Number MFCD00008720
Quantity 5 mL
Synonym 2-Phenylethyl acetate
Molecular Weight (g/mol) 164.2
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.