accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(E)

CAS RN 103-54-8

IUPAC Name: (2E)-3-phenylprop-2-en-1-yl acetate
Synonyms: cinnamyl acetate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: WJSDHUCWMSHDCR-VMPITWQZSA-N
SMILES: CC(=O)OC\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)