Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyl acetate, 99+%, Thermo Scientific™

Catalog No. AC257390010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC257390010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257390010 Supplier Thermo Scientific Chemicals Supplier No. 257390010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-54-8
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
InChI Key WJSDHUCWMSHDCR-VMPITWQZSA-N
Synonym cinnamyl acetate, 3-phenyl-2-propenyl acetate, 3-phenylallyl acetate, cinnamyl alcohol, acetate, acetic acid, cinnamyl ester, trans-cinnamyl acetate, 3-phenyl-2-propen-1-yl acetate, gamma-phenylallyl acetate, 1-acetoxy-3-phenyl-2-propene, 3-phenyl-2-propen-1-ol acetate
PubChem CID 5282110
IUPAC Name [(E)-3-phenylprop-2-enyl] acetate
SMILES CC(=O)OCC=CC1=CC=CC=C1

Specifications

Density 1.054g/mL
Assay Percent Range 99+%
Linear Formula CH3CO2CH2CH=CHC6H5
Quantity 1 L
Merck Index 15, 2302
Formula Weight 176.21
Chemical Name or Material Cinnamyl acetate, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.