accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-71-9

IUPAC Name: isocyanatobenzene
Synonyms: phenyl isocyanate phenylcarbimide carbanil PhNCO
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: DGTNSSLYPYDJGL-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)