Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Isocyanate, MP Biomedicals

Catalog No. ICN217403 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217403 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217403 Supplier MP Biomedicals, Inc Supplier No. 0521740380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 103-71-9
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
InChI Key DGTNSSLYPYDJGL-UHFFFAOYSA-N
Synonym phenyl isocyanate, phenylcarbimide, carbanil, phenyl carbonimide, benzene, isocyanato, phenylisocyanate, isocyanic acid, phenyl ester, mondur p, phnco, fenylisokyanat
PubChem CID 7672
ChEBI CHEBI:53806
IUPAC Name isocyanatobenzene
SMILES C1=CC=C(C=C1)N=C=O

Specifications

Quantity 100 g
Formula Weight 119.12
Chemical Name or Material Phenyl Isocyanate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.