accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-83-3

IUPAC Name: benzyldimethylamine
Synonyms: dimethylbenzylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: XXBDWLFCJWSEKW-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (8)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (7)