accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OHOO

CAS RN 10303-64-7

IUPAC Name: (2S)-2-hydroxy-4-methylpentanoate
Synonyms: L-2-hydroxy-4-methylvalerate
Molecular Weight (g/mol): 131.15
Molecular Formula: C6H11O3
InChi Key: LVRFTAZAXQPQHI-YFKPBYRVSA-M
SMILES: CC(C)C[C@H](O)C([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)