Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Leucic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620975 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-975 100 g
18-620-974 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-975 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1610100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: 2-Hydroxy-4-Methylpentanoic Acid, 2-Hydroxy-4-Methylvaleric Acid, 2-Hydroxyisocaproic Acid

Chemical Identifiers

CAS 10303-64-7
Molecular Weight (g/mol) 132.16 g/mol

Specifications

Percent Purity 98% Min.
Packaging Amber Bottle
Quantity 100 g
Chemical Name or Material DL-Leucic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.