accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONBrO

CAS RN 103058-87-3

IUPAC Name: 5-bromo-2-methoxypyridine-3-carbaldehyde
Synonyms: 5-bromo-2-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: VRNOWEKCASDTFG-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1C=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results