Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxypyridine-3-carboxaldehyde, 99%, Thermo Scientific™

Catalog No. AAH5006203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5006203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5006203 Supplier Thermo Scientific Chemicals Supplier No. H5006203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-methoxypyridine-3-carboxaldehyde is used as a synthetic building block. It is also used as an pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103058-87-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD05663521
InChI Key VRNOWEKCASDTFG-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy-pyridine-3-carbaldehyde, 5-bromo-2-methoxynicotinaldehyde, 5-bromo-2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 5-bromo-2-methoxy, 5-bromo-2-methoxypyridin-3-carboxaldehyde, 5-bromo-2-methoxy-3-pyridinecarboxaldehyde, pubchem9207, abbypharma ap-10-5904, 3-pyridinecarboxaldehyde,5-bromo-2-methoxy, 5-bromo-2-methoxypyridine-3-carboxaldehyde 1g
PubChem CID 1516403
IUPAC Name 5-bromo-2-methoxypyridine-3-carbaldehyde
SMILES COC1=NC=C(Br)C=C1C=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 216.04
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-methoxypyridine-3-carboxaldehyde
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.