accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NClHNNN

CAS RN 10310-21-1

IUPAC Name: 6-chloro-7H-purin-2-amine
Synonyms: 6-chloro-7H-purin-2-amine
Molecular Weight (g/mol): 169.57
Molecular Formula: C5H4ClN5
InChi Key: RYYIULNRIVUMTQ-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=C2NC=NC2=N1
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)