accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10311-61-2

IUPAC Name: N4,N4'-bis(4-methylphenyl)-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(4-methylphenyl)-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 364.49
Molecular Formula: C26H24N2
InChi Key: YUPJJGHTTBDCIK-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C(C=C2)C2=CC=C(NC3=CC=C(C)C=C3)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results