Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Di-p-tolylbenzidine 98.0+%, TCI America™
SDP

Catalog No. D35671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35671G Supplier TCI America Supplier No. D35671G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10311-61-2
Molecular Formula C26H24N2
Molecular Weight (g/mol) 364.492
InChI Key YUPJJGHTTBDCIK-UHFFFAOYSA-N
Synonym N,N′C-Bis(4-methylphenyl)benzidine
PubChem CID 18958610
IUPAC Name 4-methyl-N-[4-[4-(4-methylanilino)phenyl]phenyl]aniline
SMILES CC1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)NC4=CC=C(C=C4)C

Specifications

Melting Point 240°C
Color Yellow
Quantity 1 g
Formula Weight 364.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di-p-tolylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.