accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CONH

CAS RN 10315-03-4

IUPAC Name: 1-(4-phenylpiperidin-4-yl)ethan-1-one hydrochloridyl
Synonyms: 1-(4-phenylpiperidin-4-yl)ethanone hydrochloridyl
Molecular Weight (g/mol): 238.74
Molecular Formula: C13H17ClNO
InChi Key: ILIXCEOPJDISBX-UHFFFAOYSA-N
SMILES: [Cl].CC(=O)C1(CCNCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)