accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103260-44-2

IUPAC Name: ethyl 2-(oxan-4-yl)acetate
Synonyms: ethyl 2-(oxan-4-yl)acetate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: JLMMMEDWRUVCEW-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results