Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl tetrahydropyran-4-ylacetate, 97%, Thermo Scientific™

Catalog No. CC67423DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67423DE 5 g Amber Glass Bottle
CC67423DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67423DE Supplier Thermo Scientific Chemicals Supplier No. CC67423DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 103260-44-2
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
InChI Key JLMMMEDWRUVCEW-UHFFFAOYSA-N
Synonym ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate
PubChem CID 2773412
IUPAC Name ethyl 2-(oxan-4-yl)acetate
SMILES CCOC(=O)CC1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl tetrahydropyran-4-ylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.