accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNHNOOCH3CH3CH3

CAS RN 1032758-88-5

IUPAC Name: tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]carbamate
Synonyms: tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]carbamate
Molecular Weight (g/mol): 321.18
Molecular Formula: C15H24BN3O4
InChi Key: XICHESWFGOSKMN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results