Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-butoxycarbonylamino)pyrimidine-5-boronic acid, 97%, Thermo Scientific™

Catalog No. MAY00062DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00062DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00062DA Supplier Thermo Scientific Chemicals Supplier No. MAY00062DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1032758-88-5
Molecular Formula C15H24BN3O4
Molecular Weight (g/mol) 321.184
MDL Number MFCD09027069
InChI Key XICHESWFGOSKMN-UHFFFAOYSA-N
Synonym 2-tert-butoxycarbonylamino pyrimidine-5-boronic acid, pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, 2-tert-butoxycarbonylamino pyrimidine-5-boronic acid pinacol ester, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, 2-boc-amino pyrimidine-5-boronic acid pinacol ester, 2-tert-butoxycarbonylamino pyrimidine-5-boronic a, 2-tert-butoxycarbonyl amino pyrimidin-5-yl boronic acid pinacol ester, 2-tert-butoxycarbonylamino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine
PubChem CID 17750223
IUPAC Name tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC(=O)OC(C)(C)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 321.18
Percent Purity 97%
Chemical Name or Material 2-(tert-Butoxycarbonylamino)pyrimidine-5-boronic acid, pinacol ester
Hazard Category Warning
Hazard Statement GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.