accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOO

CAS RN 10328-92-4

IUPAC Name: 1-methyl-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Synonyms: N-methylisatoic anhydride N-methyl IA
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: KJMRWDHBVCNLTQ-UHFFFAOYSA-N
SMILES: CN1C(=O)OC(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)