Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylisatoic Anhydride (Tech.), 90%, Thermo Scientific™

Catalog No. AC156885000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156885000 500 g
AC156881000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156885000 Supplier Thermo Scientific Chemicals Supplier No. 156885000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10328-92-4
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
InChI Key KJMRWDHBVCNLTQ-UHFFFAOYSA-N
Synonym n-methylisatoic anhydride, 1-methyl-1h-benzo d 1,3 oxazine-2,4-dione, n-methyl ia, 1-methyl-2h-3,1-benzoxazine-2,4 1h-dione, 2h-3,1-benzoxazine-2,4 1h-dione, 1-methyl, n-methyl-n-carboxyanthranilic anhydride, methyl-n-carboxyanthranilic anhydride, 1-methylbenzo d 1,3-oxazine-2,4-dione, n-methyl isotoic anhydride, 1-methyl-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione
PubChem CID 25160
ChEBI CHEBI:59054
IUPAC Name 1-methyl-3,1-benzoxazine-2,4-dione
SMILES CN1C2=CC=CC=C2C(=O)OC1=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 90%
Quantity 500 g
Beilstein 27, 265
Formula Weight 177.16
Percent Purity ≥88%
Grade Technical
Physical Form Chunks
Chemical Name or Material N-Methylisatoic anhydride, tech., 90%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-714-8
RTECSNumber DM3140000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.