accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONOHNOOOCH3OCH3OH

CAS RN 103285-22-9

IUPAC Name: 1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl)-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 560.6
Molecular Formula: C31H32N2O8
InChi Key: MFDHAVFJDSRPKC-UHFFFAOYNA-N
SMILES: COC1C(O)C(COC(C2=CC=CC=C2)(C2=CC=C(OC)C=C2)C2=CC=C(OC)C=C2)OC1N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results