Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine, 98%

Catalog No. AAJ6488003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6488003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6488003 Supplier Thermo Scientific Chemicals Supplier No. J6488003
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103285-22-9 | C31H32N2O8 | 560.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103285-22-9
Molecular Formula C31H32N2O8
Molecular Weight (g/mol) 560.60
MDL Number MFCD00274123
InChI Key MFDHAVFJDSRPKC-UHFFFAOYNA-N
Synonym 5'-o-4,4'-dimethoxytrityl-2'-o-methyluridine, 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 5'-o-dmt-2'-o-methyl-uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl-uridine, dmt-2-ome-u, uridine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl, 5'-o-dmt-2'-o-methyluridine, mfdhavfjdsrpkc-yxinzvnlsa-n, 2'-o-methyl-5'-o-dmt-uridine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-o-methyluridine
PubChem CID 11039002

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Soluble in acetonitrile
Formula Weight 560.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.