accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFCl

CAS RN 1033201-89-6

IUPAC Name: 1-(3-chloro-4-fluorophenyl)piperidine
Synonyms: 1-(3-chloro-4-fluorophenyl)piperidine
Molecular Weight (g/mol): 213.68
Molecular Formula: C11H13ClFN
InChi Key: FOWVTJMHLJVBHP-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results