accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103404-57-5

IUPAC Name: heptane-1,2,3-triol
Synonyms: heptane-1,2,3-triol
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: HXYCHJFUBNTKQR-UHFFFAOYNA-N
SMILES: CCCCC(O)C(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results