Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Heptanetriol, 95%, high melting isomer, mixture of diastereoisomers

Catalog No. AC395450010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC395450010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC395450010 Supplier Thermo Scientific Chemicals Supplier No. 395450010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103404-57-5 | C7H16O3 | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-57-5
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00010517
InChI Key HXYCHJFUBNTKQR-UHFFFAOYSA-N
Synonym 1,2,3-heptanetriol, 1,2,3-trihydroxyheptane, 1-butylglycerol, 1-n-butylglycerol, 1,2,3-heptanetriol isomer t 1 m solution, 1,2,3-heptanetriol isomer h 0.5 solution, acmc-1c7bt, 1,2,3-heptanetriol solution, additive screening solution 08/fluka kit no 78374, additive screening solution 09/fluka kit no 78374
PubChem CID 124604
ChEBI CHEBI:67095
IUPAC Name heptane-1,2,3-triol
SMILES CCCCC(C(CO)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 92.0°C to 97.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH(OH)CH(OH)CH2OH
Quantity 1 g
Formula Weight 148.2
Percent Purity 95%
Chemical Name or Material 1, 2, 3-Heptanetriol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.