accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO(S)NHOHNN

CAS RN 103420-77-5

IUPAC Name: N-[(3S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl]-1H-indole-2-carboxamide
Synonyms: devazepide
Molecular Weight (g/mol): 408.46
Molecular Formula: C25H20N4O2
InChi Key: NFHRQQKPEBFUJK-HSZRJFAPSA-N
SMILES: CN1C2=CC=CC=C2C(=N[C@H](NC(=O)C2=CC3=CC=CC=C3N2)C1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)