accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 1034566-05-6

IUPAC Name: 2-bromo-1-phenyl-1H-imidazole
Synonyms: 2-bromo-1-phenylimidazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: KKPQUNLAAJJUCR-UHFFFAOYSA-N
SMILES: BrC1=NC=CN1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results