Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-bromo-1-phenyl-1H-imidazole, Thermo Scientific™

Catalog No. CC75910DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC75910DE 5 g
CC75910DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC75910DE Supplier Thermo Scientific Chemicals Supplier No. CC75910DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1034566-05-6
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
InChI Key KKPQUNLAAJJUCR-UHFFFAOYSA-N
Synonym 2-bromo-1-phenyl-1h-imidazole, 2-bromo-1-phenylimidazole
PubChem CID 45594301

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 2-bromo-1-phenyl-1H-imidazole
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.