accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10350-68-2

IUPAC Name: 2-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
Synonyms: 2-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
Molecular Weight (g/mol): 161.16
Molecular Formula: C8H7N3O
InChi Key: HIYXDTYFKNXLFO-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results