accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10350-70-6

IUPAC Name: 4-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
Synonyms: 4-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
Molecular Weight (g/mol): 161.16
Molecular Formula: C8H7N3O
InChi Key: JZIKSOZOBQKFEX-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results