accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10351-76-5

IUPAC Name: 6-methyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride
Synonyms: 6-methyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride
Molecular Weight (g/mol): 212.63
Molecular Formula: C9H9ClN2O2
InChi Key: QCJPDBNTTAJIRH-UHFFFAOYSA-N
SMILES: Cl.CC1=C(C=C2N=CNC2=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results