CAS RN 1036991-40-8
IUPAC Name:
1,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms:
1,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol):
222.1
Molecular Formula:
C11H19BN2O2
InChi Key:
ZLIQCQOFVHSHPX-UHFFFAOYSA-N
SMILES:
CN1N=CC(B2OC(C)(C)C(C)(C)O2)=C1C
1
–
1
of
1
results
1
Sigma Aldrich 1,5-Dimethyl-1H-pyrazole-4-boronic acid,pinacol ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.