accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSONHNN

CAS RN 103745-39-7

IUPAC Name: 5-(1,4-diazepane-1-sulfonyl)isoquinoline
Synonyms: fasudil 法舒地尔 fasudilum
Molecular Weight (g/mol): 291.37
Molecular Formula: C14H17N3O2S
InChi Key: NGOGFTYYXHNFQH-UHFFFAOYSA-N
SMILES: O=S(=O)(N1CCCNCC1)C1=C2C=CN=CC2=CC=C1
Form 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results