accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3NH3NH3NH3H2OH2OOHSOOOOHSOOOOHSOOOOHSOOOCe

CAS RN 10378-47-9

IUPAC Name: λ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Synonyms: ammonium cerium(IV) sulfate
Molecular Weight (g/mol): 632.53
Molecular Formula: CeH20N4O18S4
InChi Key: VCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILES: N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
Grade 
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (6)
  • (4)
Physical Form 
  • (3)
  • (6)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (6)
  • (2)
  • (4)
  • (9)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (4)
  • (6)
  • (6)
  • (4)
  • (3)
  • (6)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (6)