Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceric Ammonium Sulfate, 0.10 N (0.10 M) Solution in 1.0 N Sulfuric Acid, Spectrum™ Chemical
SDP

Catalog No. 18609613 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-613 20 L Amber Glass Bottle
18-609-610 500 mL Amber Glass Bottle
18-609-611 1 L Amber Glass Bottle
18-609-612 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-609-613 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C45620LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Ceric Ammonium Sulfate Solution
CAS 10378-47-9, 7732-18-5, 7664-93-9
Physical Form Liquid
Quantity 20 L
Grade Ungraded
Concentration or Composition (by Analyte or Components) 0.10 N (0.10 M) Solution in 1.0 N Sulfuric Acid
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.