accessibility menu, dialog, popup

UserName

Viewing profile as user:
CuONON

CAS RN 10380-28-6

IUPAC Name: copper(2+) bis(quinolin-8-olate)
Synonyms: copper(2+) bis(quinolin-8-olate)
Molecular Weight (g/mol): 351.85
Molecular Formula: C18H12CuN2O2
InChi Key: YXLXNENXOJSQEI-UHFFFAOYSA-L
SMILES: [Cu++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)