Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper 8-Hydroxyquinolate, MP Biomedicals

Catalog No. ICN213921 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213921 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213921 Supplier MP Biomedicals, Inc Supplier No. 0521392101

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10380-28-6
Molecular Formula C18H12CuN2O2
Molecular Weight (g/mol) 351.85
MDL Number MFCD00067392
InChI Key YXLXNENXOJSQEI-UHFFFAOYSA-L
Synonym cunilate, cuproquin, dokirin, fruitdo, quinolate, quinondo, milmer, copper oxinate, copper oxine, copper-oxine
PubChem CID 56840841
ChEBI CHEBI:82028
IUPAC Name copper(2+) bis(quinolin-8-olate)
SMILES [Cu++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1

Specifications

Quantity 100 g
Formula Weight 351.86
Chemical Name or Material Copper 8-Hydroxyquinolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.