accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10382-52-2

H3CCH3OHONHSNOOHN

CAS RN 10382-52-2

IUPAC Name: 3-methyl-2-{[(2-nitrophenyl)sulfanyl]amino}pentanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 3-methyl-2-{[(2-nitrophenyl)sulfanyl]amino}pentanoic acid; dicha
Molecular Weight (g/mol): 465.65
Molecular Formula: C24H39N3O4S
InChi Key: IBYYSIWLIRUKII-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CCC(C)C(NSC1=CC=CC=C1[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)