Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-2-Nitrophenylsulfenyl-L-isoleucine Dicyclohexylammonium Salt 98.0+%, TCI America™
SDP

Catalog No. N02401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N02401G Supplier TCI America Supplier No. N02401G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10382-52-2
Molecular Formula C24H39N3O4S
Molecular Weight (g/mol) 465.653
MDL Number MFCD00038495
InChI Key IBYYSIWLIRUKII-RWHJDYSMSA-N
Synonym N-(2-Nitrophenylthio)-L-isoleucine Dicyclohexylammonium Salt, N-Nps-L-isoleucine Dicyclohexylammonium Salt, Nps-Ile-OH.DCHA
PubChem CID 91659100
IUPAC Name N-cyclohexylcyclohexanamine;(2S,3S)-3-methyl-2-[(2-nitrophenyl)sulfanylamino]pentanoic acid
SMILES CCC(C)C(C(=O)O)NSC1=CC=CC=C1[N+](=O)[O-].C1CCC(CC1)NC2CCCCC2

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 465.65
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-2-Nitrophenylsulfenyl-L-isoleucine Dicyclohexylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.