accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103980-68-3

CH3CH3NHNO

CAS RN 103980-68-3

IUPAC Name: 6-methyl-5-propyl-1,4-dihydropyrimidin-4-one
Synonyms: 6-methyl-5-propyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 152.2
Molecular Formula: C8H12N2O
InChi Key: UAJIFHDSCLQUON-UHFFFAOYSA-N
SMILES: CCCC1=C(C)NC=NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results