accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-20-1

IUPAC Name: 4-(4-methoxyphenyl)butan-2-one
Synonyms: methoxyphenylbutanone
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: PCBSXBYCASFXTM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCC(C)=O)C=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results