Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methoxyphenyl)-2-butanone 97.0+%, TCI America™
SDP

Catalog No. M075625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M075625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M075625ML Supplier TCI America Supplier No. M075625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104-20-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008791
InChI Key PCBSXBYCASFXTM-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
PubChem CID 61007
IUPAC Name 4-(4-methoxyphenyl)butan-2-one
SMILES COC1=CC=C(CCC(C)=O)C=C1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 177°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone
EINECSNumber (3)-2677
RTECSNumber EL9085500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.