accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-74-5

IUPAC Name: 1-dodecylpyridin-1-ium chloride
Synonyms: laurylpyridinium chloride
Molecular Weight (g/mol): 283.88
Molecular Formula: C17H30ClN
InChi Key: GKQHIYSTBXDYNQ-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results