Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecylpyridinium Chloride 98.0+%, TCI America™
SDP

Catalog No. D0995500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0995500G 500 g
D099525G 25 g
D0995100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0995500G Supplier TCI America Supplier No. D0995500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104-74-5 | C17H30ClN | 283.884 g/mol

Chemical Identifiers

CAS 104-74-5
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.884
MDL Number MFCD00011987
InChI Key GKQHIYSTBXDYNQ-UHFFFAOYSA-M
Synonym 1-Laurylpyridinium Chloride
PubChem CID 7717
IUPAC Name 1-dodecylpyridin-1-ium;chloride
SMILES CCCCCCCCCCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Quantity 500 g
Formula Weight 283.88
Percent Purity ≥98.0% (HPLC)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecylpyridinium Chloride
RTECSNumber UU4017070
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.