CAS RN 1040281-97-7
IUPAC Name:
2-(3-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-(3-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
244.54
Molecular Formula:
C10H14BClO2S
InChi Key:
UAAAZOFUOOBDBI-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(Cl)C=CS1
1
–
1
of
1
results
1
Sigma Aldrich 3-Chlorothiophene-2-boronic acid pinacol ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.