accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNONFFF

CAS RN 104048-92-2

IUPAC Name: 6-(trifluoromethyl)-1,2-dihydropyrimidin-2-one
Synonyms: 4-(trifluoromethyl)-3H-pyrimidin-2-one
Molecular Weight (g/mol): 164.09
Molecular Formula: C5H3F3N2O
InChi Key: WCEBFEVZTGLOHC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=NC(=O)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results