CAS RN 104076-39-3
CAS RN 104076-39-3
IUPAC Name:
N-(3-{3-[(piperidin-1-yl)methyl]phenoxy}propyl)-1,3-benzothiazol-2-amine; bis(formic acid); bis(prop-2-enoic acid)
Synonyms:
bis(acrylic acid); bis(formic acid); zolantidine
Molecular Weight (g/mol):
617.71
Molecular Formula:
C30H39N3O9S
InChi Key:
LUWINHZYJPMGDV-UHFFFAOYSA-N
SMILES:
OC=O.OC=O.OC(=O)C=C.OC(=O)C=C.C(CNC1=NC2=CC=CC=C2S1)COC1=CC=CC(CN2CCCCC2)=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Zolantidine dimaleate
CAS: 104076-39-3 Molecular Formula: C30H39N3O9S